missing translation for 'onlineSavingsMsg'
Learn More

ERP29 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Product Code. 18354036 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18354036 20 μL 20µL
18378784 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18354036 Supplier Novus Biologicals Supplier No. NBP30307220UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ERP29 Polyclonal specifically detects ERP29 in Human, Mouse, Rat samples. It is validated for Western Blot, Knockout Validated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERP29
Applications Western Blot, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias C12orf8chromosome 12 open reading frame 8, endoplasmic reticulum lumenal protein ERp28, endoplasmic reticulum protein 29, Endoplasmic reticulum resident protein 28, endoplasmic reticulum resident protein 29, ERp28Endoplasmic reticulum resident protein 31, ERp29, ERp31ERP28, PDIA9, PDI-DB, protein disulfide isomerase family A, member 9
Host Species Rabbit
Immunogen A synthetic peptide of human ERP29.
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Membrane Trafficking and Chaperones, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10961
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.