missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERO1LB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18312-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody has no cross reaction to ERO1L.
ERO1LB is essential oxidoreductase that oxidizes proteins in the endoplasmic reticulum to produce disulfide bonds. ERO1LB acts by oxidizing directly P4HB/PDI isomerase through a direct disulfide exchange. ERO1LB does not act as a direct oxidant of folding substrate, but relies on P4HB/PDI to transfer oxidizing equivalent.Specifications
| ERO1LB | |
| Polyclonal | |
| Unconjugated | |
| ERO1B | |
| ERO1 L beta, ERO1 like beta (S. cerevisiae), ERO1 like protein beta, ERO1LB, Oxidoreductin 1 L beta | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 364755, 56605, 67475 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q86YB8 | |
| Ero1b, Ero1lb | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction