missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ERO1LB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA525142
This item is not returnable.
View return policy
Description
ERO1LB Polyclonal Antibody for Flow, IHC (P), Western Blot
ERO1LB is essential oxidoreductase that oxidizes proteins in the endoplasmic reticulum to produce disulfide bonds. ERO1LB acts by oxidizing directly P4HB/PDI isomerase through a direct disulfide exchange. ERO1LB does not act as a direct oxidant of folding substrate, but relies on P4HB/PDI to transfer oxidizing equivalent.
Specifications
| ERO1LB | |
| Polyclonal | |
| Unconjugated | |
| Ero1b | |
| 1300013B24Rik; 1700065B09Rik; AI447560; endoplasmic oxidoreductase 1 beta; endoplasmic oxidoreductin-1-like protein B; endoplasmic reticulum oxidoreductase 1 beta; endoplasmic reticulum oxidoreductase beta; Endoplasmic reticulum oxidoreductin-1-like protein B; Ero1b; Ero1beta; ero1-beta; ERO1LB; ERO1-L-beta; ERO1-like beta; ERO1-like beta (S. cerevisiae); ERO1-like protein beta; Oxidoreductin-1-L-beta; RGD1563548 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 56605, 67475 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q86YB8, Q8R2E9 | |
| Ero1b, Ero1lb | |
| KLH conjugated synthetic peptide between 266-293 amino acids from the C-terminal region of human ERO1LB. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction