missing translation for 'onlineSavingsMsg'
Learn More

ERO1L Antibody (V23P3C9*H2), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18724411 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18724411 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18724411 Supplier Novus Biologicals Supplier No. NBP250388MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ERO1L Monoclonal antibody specifically detects ERO1L in Human, Mouse, Rat, Porcine, Canine, Equine, Sheep samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERO1L
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone V23P3C9*H2
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias EC 1.8.4, EC 1.8.4.-, Endoplasmic oxidoreductin-1-like protein, ERO1 (S. cerevisiae)-like, ERO1A, ERO1-alpha, ERO1-L, ERO1-L-alpha, ERO1-like (S. cerevisiae), ERO1-like protein alpha, oxidoreductin-1-L-alpha
Host Species Mouse
Immunogen Peptide Sequence - KLIANMPESG (peptide immunogen is identical in rat (Rattus norvegicus), mouse (Mus musculus), sheep (Ovis aries), bovine (Bos taurus), canine (Canis familiaris), porcine (Sus scrofa) and equine (Equus caballus)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, ER Markers, Hypoxia, Membrane Trafficking and Chaperones, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 30001
Target Species Human, Mouse, Rat, Pig, Canine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.