missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERO1L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£188.00 - £422.00
Specifications
| Antigen | ERO1L |
|---|---|
| Concentration | 2 mg/mL |
| Dilution | Western Blot 1:1000-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18074884
|
Novus Biologicals
NB100-2525SS |
0.025 mL |
£188.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18715783
|
Novus Biologicals
NB100-2525 |
0.1 mL |
£422.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ERO1L Polyclonal specifically detects ERO1L in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| ERO1L | |
| Western Blot 1:1000-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Polyclonal | |
| Rabbit | |
| Cancer, ER Markers, Hypoxia, Membrane Trafficking and Chaperones, Signal Transduction | |
| PBS and 1.0% BSA with No Preservative | |
| EC 1.8.4, EC 1.8.4.-, Endoplasmic oxidoreductin-1-like protein, ERO1 (S. cerevisiae)-like, ERO1A, ERO1-alpha, ERO1-L, ERO1-L-alpha, ERO1-like (S. cerevisiae), ERO1-like protein alpha, oxidoreductin-1-L-alpha | |
| ERO1L | |
| IgG | |
| Affinity Purified | |
| 57 kDa |
| 2 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| Q96HE7 | |
| 30001 | |
| A synthetic peptide made to the N-terminal region of human ERO1L [UniProt Q96HE7] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title