missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERManI Antibody (3C2) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80726
This item is not returnable.
View return policy
Description
ERManI Monoclonal specifically detects ERManI in Human, Mouse (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated.
Specifications
| ERManI | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot : 1:500-1:1000, ELISA : 1:1000, Immunohistochemistry : 1:200, Immunocytochemistry/Immunofluorescence : 1:100, Immunoprecipitation : 1:10-1:100, Immunohistochemistry-Paraffin : 1:200, Knockout Validated reported in scientific literature (PMID 26205822) | |
| alpha 1,2-mannosidase, EC 3.2.1.113, endoplasmic reticulum alpha-mannosidase 1, endoplasmic reticulum mannosyl-oligosaccharide 1,2-alpha-mannosidase, endoplasmic reticulum mannosyl-oligosaccharide 1,2-alpha-mannosidase 1, ER alpha 1,2-mannosidase, ER alpha-1,2-mannosidase, ER ManI, ER mannosidase 1, ERMan1, Man9GlcNAc2-specific processing alpha-mannosidase, Man9GlcNAc2-specific-processing alpha-mannosidase, MANA-ER, Mannosidase alpha class 1B member 1, mannosidase, alpha, class 1B, member 1, MRT15, RP11-350O14.2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human. Does not cross react with mouse. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 3C2 | |
| Unconjugated | |
| Tris-Glycine (pH 7.5), 0.15M NaCl | |
| MAN1B1 | |
| Full length human recombinant ERManI [Swiss-Prot# Q9UKM7] | |
| 0.1 mL | |
| Cellular Markers | |
| 11253 | |
| Human, Mouse (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction