missing translation for 'onlineSavingsMsg'
Learn More

Erlotinib, TRC

Product Code. 30387706
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387706 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387706 Supplier Toronto Research Chemicals Supplier No. TRCE62500825G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Erlotinib
CAS 183321-74-6
Molecular Formula C22 H23 N3 O4
Quantity 25 g
Synonym Erlotinib, 4-Quinazolinamine, N-(3-ethynylphenyl)-6,7-bis(2-methoxyethoxy)-, N-(3-Ethynylphenyl)-6,7-bis(2-methoxyethoxy)-4-quinazolinamine, 4-[(3-Ethynylphenyl)amino]-6,7-bis(2-methoxyethoxy)quinazoline, NSC 718781, OSI 744, R 1415
InChI Formula InChI=1S/C22H23N3O4/c1-4-16-6-5-7-17(12-16)25-22-18-13-20(28-10-8-26-2)21(29-11-9-27-3)14-19(18)23-15-24-22/h1,5-7,12-15H,8-11H2,2-3H3,(H,23,24,25)
SMILES COCCOc1cc2ncnc(Nc3cccc(c3)C#C)c2cc1OCCOC
IUPAC Name N-(3-ethynylphenyl)-6,7-bis(2-methoxyethoxy)quinazolin-4-amine
Molecular Weight (g/mol) 393.44
Formula Weight 393.1689
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.