missing translation for 'onlineSavingsMsg'
Learn More

Erlotinib-d6 Hydrochloride, TRC

Product Code. 30386801
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386801 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30386801 Supplier Toronto Research Chemicals Supplier No. TRCE62500210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Erlotinib bis-methoxy-d6 hydrochloride
CAS 1189953-78-3
Purity Grade Notes HPLC
Molecular Formula C22 2H6 H17 N3 O4 . Cl H
Quantity 10 mg
Synonym N-(3-Ethynylphenyl)-6,7-bis[2-(trideuteriomethoxy)ethoxy]quinazolin-4-amine hydrochloride, N-(3-Ethynylphenyl)-6,7-bis[2-(methoxy-d3)ethoxy]-4-quinazolinamine hydrochloride, Erlotinib-d6 hydrochloride, Erlotinib-D6 hydrochloride (bis(methoxy-D3))
InChI Formula InChI=1S/C22H23N3O4.ClH/c1-4-16-6-5-7-17(12-16)25-22-18-13-20(28-10-8-26-2)21(29-11-9-27-3)14-19(18)23-15-24-22;/h1,5-7,12-15H,8-11H2,2-3H3,(H,23,24,25);1H/i2D3,3D3;
SMILES Cl.[2H]C([2H])([2H])OCCOc1cc2ncnc(Nc3cccc(c3)C#C)c2cc1OCCOC([2H])([2H])[2H]
IUPAC Name N-(3-ethynylphenyl)-6,7-bis[2-(trideuteriomethoxy)ethoxy]quinazolin-4-amine;hydrochloride
Molecular Weight (g/mol) 435.93
Formula Weight 435.1832
Percent Purity >95
Show More Show Less
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.