missing translation for 'onlineSavingsMsg'
Learn More

ERLIN1 Antibody (OTI4C11) [DyLight 550], Novus Biologicals™

Product Code. 30687993 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30687993 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30687993 Supplier Novus Biologicals Supplier No. NBP344917R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ERLIN1 Monoclonal antibody specifically detects ERLIN1 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERLIN1
Applications Western Blot
Classification Monoclonal
Clone OTI4C11
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias Band_7 23-211 Keo4 (Interim) similar to C.elegans protein C42C1.9, C10orf69, chromosome 10 open reading frame 69, Endoplasmic reticulum lipid raft-associated protein 1, ER lipid raft associated 1, Erlin-1, KE04SPFH domain-containing protein 1, KEO4, Protein KE04, SPFH domain family, member 1, SPFH1, Stomatin-prohibitin-flotillin-HflC/K domain-containing protein 1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 107-346 of human ERLIN1(NP_006450) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10613
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.