Learn More
Karl Hecht™ Erlenmeyer Flask with Hollow NS Glass Stopper
Product Code. 10728662
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
Closure Size:
NS 14/23
NS 19/26
NS 29/32
Material:
Borosilicate Glass (3.3)
Borosilicate Glass, Duran Glass
Unit Size:
Each
Product Code. 10728662
Brand: Karl Hecht™ 41918042
Only null left
Add to basket
Add to basket
Conical, flat-bottomed flask well suited to mixing by swirling. Karl Hecht™ Erlenmeyer Flask with Hollow NS Glass Stopper is constructed of durable Duran™ borosilicate 3.3 glass.
  • Durable Duran™ borosilicate 3.3 glass.
  • Graduated.
  • Reinforced ground neck marked with standard ground size.
  • Hollow NS glass stopper.
Specifications
Closure Material Glass
Capacity (Metric) 500 mL
Material Borosilicate Glass (3.3)
Standard Taper Joint NS 29/32
Graduated Yes
Closure Type NS Stopper
Closure Size NS 29/32