missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERLEC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£231.00 - £444.00
Specifications
| Antigen | ERLEC1 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18402741
|
Novus Biologicals
NBP1-86802-25ul |
25 μL |
£231.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18039313
|
Novus Biologicals
NBP1-86802 |
0.1 mL |
£444.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ERLEC1 Polyclonal specifically detects ERLEC1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ERLEC1 | |
| Polyclonal | |
| Rabbit | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 27248 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:PFKPLTLRQLEQQEEILRVPFRRNKEEDLQSTKEERFPAIHKSIAIGSQPVLTVGTTHISKLTDDQLIKEFLSGSYCFRGGVGWWKYE | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Unconjugated | |
| RUO | |
| C20orf30, C2orf30, CL24936, CL25084, endoplasmic reticulum lectin 1, ER lectin, Erlectin, erlectin 1, XTP3-B, XTP3TPBchromosome 2 open reading frame 30, XTP3-transactivated gene B protein, XTP3-transactivated protein B | |
| ERLEC1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title