Learn More
3M™ Versaflo™ TR-600 PAPR Accessory, Battery Charger Kit
Product Code. 16100841
Change view
Click to view available options
Model:
TR-641E
TR-641UK
TR-644E
TR-644UK
Product Type:
Four Station Battery Charger Kit
Single Station Battery Charger Kit
Unit Size:
Each
Includes:
Four TR-640 Cradles, a Baseplate, Power Supply and Lead
TR-640 Cradle, Power Supply and Lead
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Product Type unitSize Includes
16100841 TR-644UK Four Station Battery Charger Kit Each -
15509265 TR-641E Single Station Battery Charger Kit Each TR-640 Cradle, Power Supply and Lead
15519265 TR-644E Four Station Battery Charger Kit Each Four TR-640 Cradles, a Baseplate, Power Supply and Lead
15710058 TR-641UK Single Station Battery Charger Kit Each TR-640 Cradle, Power Supply and Lead
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 16100841 Supplier 3M™ Supplier No. 7100020282
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
3M Versaflo™ TR-600 PAPR Accessory, Battery Charger Kit is used with 3M™ Versaflo™ TR-600 series air purifying device.
  • Charges TR-630 standard battery or TR-632 high capacity battery in less than 4 hours
  • Batteries may be stored on the charger, fully charged, ready for immediate use
TRUSTED_SUSTAINABILITY
Specifications
Product Type Four Station Battery Charger Kit
For Use With (Equipment) 3M Versaflo Powered Air Turbo TR-600 Series
Model TR-644UK