Learn More
3M™ Versaflo™ TR-600 PAPR Accessory, Battery Charger Kit
3M Versaflo™ TR-600 PAPR Accessory, Battery Charger Kit is used with 3M™ Versaflo™ TR-600 series air purifying device.
Brand: 3M™ 7100021098
Code : NEW
Additional Details : Weight : 0.63300kg
Description
- Charges TR-630 standard battery or TR-632 high capacity battery in less than 4 hours
- Batteries may be stored on the charger, fully charged, ready for immediate use
Specifications
Single Station Battery Charger Kit | |
3M Versaflo Powered Air Turbo TR-600 Series | |
TR-640 Cradle, Power Supply and Lead | |
TR-641UK |