Learn More
3M™ Versaflo™ TR-600 PAPR Accessory, Battery Charger Kit
3M Versaflo™ TR-600 PAPR Accessory, Battery Charger Kit is used with 3M™ Versaflo™ TR-600 series air purifying device.
Brand:  3M™ 7000144505
789.00 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 0.63000kg Transport : UN number : 3481 Chem class : 9
 View more versions of this product
Product Code. 15519265
  • £1045.00 / Each
Estimated Shipment: 24-06-2024
to see stock.
Add to basket
Description
Description
  • Charges TR-630 standard battery or TR-632 high capacity battery in less than 4 hours
  • Batteries may be stored on the charger, fully charged, ready for immediate use
Specifications
Specifications
Four Station Battery Charger Kit
3M Versaflo Powered Air Turbo TR-600 Series
Four TR-640 Cradles, a Baseplate, Power Supply and Lead
TR-644E