missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERK3/MAPK6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£178.00 - £366.00
Specifications
| Antigen | ERK3/MAPK6 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18318476
|
Novus Biologicals
NBP3-03701-20UL |
20 μL |
£178.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18303414
|
Novus Biologicals
NBP3-03701-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ERK3/MAPK6 Polyclonal specifically detects ERK3/MAPK6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.Spécification
| ERK3/MAPK6 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DKFZp686F03189, EC 2.7.11, EC 2.7.11.24, ERK-3, ERK3p97-MAPK, Extracellular signal-regulated kinase 3, extracellular signal-regulated kinase, p97, HsT17250, MAP kinase 6, MAP kinase isoform p97, MAPK 6, mitogen-activated protein kinase 6, p97MAPK, PRKM6, protein kinase, mitogen-activated 5, protein kinase, mitogen-activated 6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 372-721 of human ERK3/MAPK6 (NP_002739.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunofluorescence, KnockDown | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Cellular Markers, Hypoxia, Phospho Specific, Signal Transduction, Stem Cell Markers | |
| PBS with 50% glycerol, pH7.3. | |
| 5597 | |
| IgG | |
| Affinity purified |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit