missing translation for 'onlineSavingsMsg'
Learn More

ERK3/MAPK6 Antibody [HRP], Novus Biologicals™

Product Code. 30660811 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30660811 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30660811 Supplier Novus Biologicals Supplier No. AF3196H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

ERK3/MAPK6 Polyclonal antibody specifically detects ERK3/MAPK6 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERK3/MAPK6
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias DKFZp686F03189, EC 2.7.11, EC 2.7.11.24, ERK-3, ERK3p97-MAPK, Extracellular signal-regulated kinase 3, extracellular signal-regulated kinase, p97, HsT17250, MAP kinase 6, MAP kinase isoform p97, MAPK 6, mitogen-activated protein kinase 6, p97MAPK, PRKM6, protein kinase, mitogen-activated 5, protein kinase, mitogen-activated 6
Host Species Goat
Immunogen E. coli-derived recombinant human ERK3, Met344-Asn721, Accession # Q16659
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Cellular Markers, Hypoxia, Phospho Specific, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5597
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.