missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ERK2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517710
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
MAPK1 (ERK2) is a serine/threonine kinase that acts as an important activator of p90 RSK, MSK, ELK1, and Stat3, and is involved in the MAPK pathway. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. The activation of this kinase requires its phosphorylation by upstream kinases. Upon activation, this kinase translocates to the nucleus of the stimulated cells, where it phosphorylates nuclear targets.
Specifications
| ERK2 | |
| Polyclonal | |
| Unconjugated | |
| MAPK1 | |
| 9030612K14Rik; AA407128; AU018647; C78273; ERK; ERK2; ERK-2; ERT1; extracellular signal-regulated kinase 2; extracellular signal-regulated kinase-2; extracellular-signal-regulated kinase 2; MAP kinase 1; MAP kinase 2; MAP kinase isoform p42; Mapk; MAPK 1; MAPK 2; Mapk1; MAPK2; mitogen activated protein kinase 1; mitogen-activated protein kinase 1; Mitogen-activated protein kinase 2; p38; p40; p41; p41mapk; P42MAPK; p42-MAPK; Prkm1; PRKM2; protein tyrosine kinase ERK2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116590, 26413, 5594 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 92 μg/mL | |
| 0.01M HEPES with 50% glycerol, 100μg/mL BSA, 0.15M NaCl and no preservative; pH 7.5 | |
| P28482, P63085, P63086 | |
| MAPK1 | |
| Synthetic peptide corresponding to human p42 MAP kinase (Erk2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion