missing translation for 'onlineSavingsMsg'
Learn More

ERK1/ERK2 Antibody (631122) [CoraFluor™ 1], Novus Biologicals™

Product Code. 30666940 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30666940 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30666940 Supplier Novus Biologicals Supplier No. FAB15761CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ERK1/ERK2 Monoclonal antibody specifically detects ERK1/ERK2 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERK1/ERK2
Applications Western Blot
Classification Monoclonal
Clone 631122
Conjugate CoraFluor 1
Formulation PBS
Gene Alias ERK1, ERK-1, ERT2, Extracellular signal-regulated kinase 1, extracellular signal-related kinase 1, HS44KDAP, HUMKER1A, Insulin-stimulated MAP2 kinase, MAPK 1, Microtubule-associated protein 2 kinase, mitogen-activated protein kinase 3, p44erk1, p44-ERK1, p44mapk, p44-MAPK, PRKM3
Host Species Mouse
Immunogen Human ERK1 synthetic peptide, KKISPFEHQTYCQRTLR, Accession # P27361 (ERK1) & P28482 (ERK2)
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5595
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.