missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ERK1 Monoclonal antibody specifically detects ERK1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Dilution | Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence |
| Host Species | Mouse |
| Immunogen | Recombinant human ERK1 protein (1-137aa) purified from E. coli |
| Quantity | 0.1 mL |
| Regulatory Status | RUO |
| Research Discipline | Angiogenesis, Breast Cancer, Cancer, Cell Cycle and Replication, Diabetes Research, Hypoxia, MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?