missing translation for 'onlineSavingsMsg'
Learn More

ERK1 Antibody (1A2), DyLight 594, Novus Biologicals™

Product Code. 18423336 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423336 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423336 Supplier Novus Biologicals Supplier No. NBP174032DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ERK1 Monoclonal antibody specifically detects ERK1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERK1
Applications Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 1A2
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.11, ERK-1, ERK1p44-MAPK, ERT2, Extracellular signal-regulated kinase 1, extracellular signal-related kinase 1, HS44KDAP, HUMKER1A, Insulin-stimulated MAP2 kinase, MAP kinase 1, MAP kinase 3, MAP kinase isoform p44, MAPK 1, MAPK 3, MGC20180, Microtubule-associated protein 2 kinase, Mitogen-activated protein kinase 1, mitogen-activated protein kinase 3, p44erk1, p44-ERK1, p44mapk, PRKM3EC 2.7.11.24
Host Species Mouse
Immunogen Recombinant human ERK1 protein (1-137aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Breast Cancer, Cancer, Cell Cycle and Replication, Diabetes Research, Hypoxia, MAP Kinase Signaling, mTOR Pathway, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5595
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.