missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERK1/2 Antibody (SB46b), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-28628
This item is not returnable.
View return policy
Description
ERK1/2 Monoclonal specifically detects ERK1/2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| ERK1/2 | |
| Monoclonal | |
| HRP | |
| EC 2.7.11, ERK-1, ERK1p44-MAPK, ERT2, Extracellular signal-regulated kinase 1, extracellular signal-related kinase 1, HS44KDAP, HUMKER1A, Insulin-stimulated MAP2 kinase, MAP kinase 1, MAP kinase 3, MAP kinase isoform p44, MAPK 1, MAPK 3, MGC20180, Microtubule-associated protein 2 kinase, Mitogen-activated protein kinase 1, mitogen-activated protein kinase 3, p44erk1, p44-ERK1, p44mapk, PRKM3EC 2.7.11.24 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 5595 | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| SB46b | |
| Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500 | |
| MAPK3 | |
| Human recombinant Erk1 and Erk 2 | |
| 1 mL | |
| Primary | |
| Reacts with human Erk 1 and human Erk2 | |
| Store at 4C. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction