missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Eriochrome∣r Blue Black B

Product Code. 15485639
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15485639 100 g 100g
15495639 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15485639 Supplier Thermo Scientific Chemicals Supplier No. J63396.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3564-14-5 | C20H13N2NaO5S | 416.38 g/mol

Eriochrome is to do titration indicator. Dubbed in aqueous solution or alcohol-soluble when used for the determination of calcium, magnesium, manganese, zinc and other metal ions, measured water hardness.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Eriochrome is to do titration indicator. Dubbed in aqueous solution or alcohol-soluble when used for the determination of calcium, magnesium, manganese, zinc and other metal ions, measured water hardness.

Solubility
Soluble in water(20 g/L (20 C).

Notes
Eriochrome is a registered trademark of Ciba Specialty Chemicals Corp.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3564-14-5
Molecular Formula C20H13N2NaO5S
Molecular Weight (g/mol) 416.38
MDL Number MFCD00003934
InChI Key YRFCCVCSEVISHI-GXTSIBQPSA-M
Synonym C.I. 14640; Mordant Black 3
PubChem CID 23722702
SMILES [Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC=CC=C12)S([O-])(=O)=O

Specifications

Solubility Information Soluble in water(20g/L (20 C).
IUPAC Name sodium;4-[(1-hydroxynaphthalen-2-yl)hydrazinylidene]-3-oxonaphthalene-1-sulfonate
Formula Weight 416.39
Color Black to Brown
Quantity 100 g
Physical Form Powder
Chemical Name or Material Eriochrome™ Blue Black B

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.