missing translation for 'onlineSavingsMsg'
Learn More

Eriochrome™ Black T, Indicator, Reag. Ph. Eur. for metal titration, Honeywell Fluka™

Product Code. 15623850 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass Bottle
Unit Size:
100g
1kg
25g
This item is not returnable. View return policy

Product Code. 15623850

Brand: Honeywell-Fluka 327511KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Indicator, Reag. Ph. Eur. for metal titration

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1787-61-7
Molecular Formula C20H12N3NaO7S
Molecular Weight (g/mol) 461.38
MDL Number MFCD00003935
InChI Key JHUJLRKQZAPSDP-GXTSIBQPSA-M
PubChem CID 87355429
SMILES [Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC(=CC=C12)[N+]([O-])=O)S([O-])(=O)=O

Specifications

UN Number NONH for all modes of transport
Beilstein 4121162
IUPAC Name (4Z)-4-[(1-hydroxynaphthalen-2-yl)hydrazinylidene]-7-nitro-3-oxonaphthalene-1-sulfonic acid;sodium
Formula Weight 461.38g/mol
Packaging Glass Bottle
Suitability Complies for indicator
Quantity 1 kg
Chemical Name or Material Eriochrome™ Black T, Reag. Ph. Eur.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.