missing translation for 'onlineSavingsMsg'
Learn More

Erio Green B (Technical Grade), TRC

Product Code. 30387775
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387775 100 mg 100mg
30380752 50 mg -
30388071 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387775 Supplier Toronto Research Chemicals Supplier No. TRCE600093100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12768-78-4
Molecular Formula C31 H33 N2 O6 S2 . Na
Molecular Weight (g/mol) 616.72
Synonym 2,7-Naphthalenedisulfonic acid, 4-[(4-aminophenyl)(4-imino-2,5-cyclohexadien-1-ylidene)methyl]-, N-Et- or N-Me-quaternized, hydroxide, inner salt, sodium salt, Acid Green 16, Acid Green V, Acid Green V Naphthaline, Acid Green VS, Acid Green Zh, Acid Naphthalene Green V, Acid Pure Green V, Amido Green V, Anadurm Green A-V, BRYacid Green V, Best Acid Naphthalene Green V, Borunil Green A-FG, Colocid Green V, Covalene Brilliant Green G, Daiwa Green 70, Dinacid Green V, Duasyn Acid Green V, Duasyn Acid Green VS, Dyacid Green B, Dycosacid Green VS, Erio Green B, Everlan Green EV, Hispacid Brilliant Green SA 2G, Ichoacid Green V, Indacid Green V, Kemacid Green V, Lecotan Green J, Leoctan Green J, Libacid Green LV, Lissamine Green V, Lissamine Green V 200, Merantine Green V, Naphthalene Green V, Naphthalene Green VS, Naphthalene Green VSC, Naphthalene Green Y, Naphthazine Brilliant Green G, Pacid Green V, Ratna Acid Green V, Ravi Acid Green V, Sandolan Brilliant Green E-B, Sandolan Brilliant Green E-BI, Simacid Acid Green, Triacid Pure Green V, Triacor Green VT, Vicoacid Green V
IUPAC Name sodium;4-[[4-(diethylamino)phenyl]-(4-diethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]naphthalene-2,7-disulfonate
SMILES [Na+].CCN(CC)c1ccc(cc1)C(=C2C=CC(=[N+](CC)CC)C=C2)c3cc(cc4cc(ccc34)S(=O)(=O)[O-])S(=O)(=O)[O-]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C31H34N2O6S2.Na/c1-5-32(6-2)25-13-9-22(10-14-25)31(23-11-15-26(16-12-23)33(7-3)8-4)30-21-28(41(37,38)39)20-24-19-27(40(34,35)36)17-18-29(24)30;/h9-21H,5-8H2,1-4H3,(H-,34,35,36,37,38,39);/q;+1/p-1
Formula Weight 616.1678
Chemical Name or Material Acid Green 16
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.