missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERH Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18856AF532
This item is not returnable.
View return policy
Description
ERH Polyclonal antibody specifically detects ERH in Human samples. It is validated for ELISA
Specifications
| ERH | |
| Polyclonal | |
| ELISA | |
| DROER, enhancer of rudimentary (Drosophila) homolog, enhancer of rudimentary homolog, enhancer of rudimentary homolog (Drosophila), FLJ27340 | |
| Produced in rabbits immunized with purified, recombinant Human ERH (Uniprot # P84090; Met1-Lys104) | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 2079 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction