Learn More
Mettler Toledo™ ErgoSens
Shop All Mettler Toledo Products
Click to view available options
:
ErgoSens
Specifications
Specifications
| For Use With (Equipment) | XP and XS Balances |
| For Use With (Application) | Perform hands-free operations such as taring, zeroing or printing |