missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ERGIC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531996
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (96%), Rat (97%), Zebrafish (86%), Xenopus laevis (83%), Rabbit (97%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ERGIC3 is possible role in transport between endoplasmic reticulum and Golgi.
Specifications
| ERGIC3 | |
| Polyclonal | |
| Unconjugated | |
| ERGIC3 | |
| 2310015B14Rik; AV318804; C20orf47; CGI-54; D2Ucla1; dJ477O4.2; Endoplasmic reticulum-Golgi intermediate compartment protein 3; endoplasmic reticulum-localized protein ERp43; ERGIC and golgi 3; ERGIC3; ERV46; NY-BR-84; PRO0989; Sdbcag84; serologically defined breast cancer antigen 84; serologically defined breast cancer antigen NY-BR-84; Serologically defined breast cancer antigen NY-BR-84 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51614 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.65 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9Y282 | |
| ERGIC3 | |
| Recombinant protein encompassing a sequence within the center region of human ERGIC3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction