missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ERGIC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100505
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ERGIC2.
ERGIC2, or PTX1, is a ubiquitously expressed nuclear protein that is downregulated in prostate carcinoma (Kwok et al., 2001).
Specifications
| ERGIC2 | |
| Polyclonal | |
| Unconjugated | |
| ERGIC2 | |
| 1200009B18Rik; 4930572C01Rik; AA408438; cd002; CD14 protein; CDA14; Endoplasmic reticulum-Golgi intermediate compartment protein 2; ERGIC and golgi 2; ERGIC2; ERV41; PTX1; PTX1 protein; RGD1310606 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 51290 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96RQ1 | |
| ERGIC2 | |
| A synthesized peptide derived from human ERGIC2(Accession Q96RQ1), corresponding to amino acid residues F338-H377. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction