missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ERGIC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577205
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
ERGIC2, or PTX1, is a ubiquitously expressed nuclear protein that is downregulated in prostate carcinoma (Kwok et al., 2001).
Specifications
| ERGIC2 | |
| Polyclonal | |
| Unconjugated | |
| ERGIC2 | |
| 1200009B18Rik; 4930572C01Rik; AA408438; cd002; CD14 protein; CDA14; Endoplasmic reticulum-Golgi intermediate compartment protein 2; ERGIC and golgi 2; ERGIC2; ERV41; PTX1; PTX1 protein; RGD1310606 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51290, 67456 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96RQ1, Q9CR89 | |
| ERGIC2 | |
| Recombinant full length Human ERGIC2. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction