Learn More
Invitrogen™ ERG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534810
Description
Recommended positive controls: A431, H1299, HeLa, Molt-4, Raji, mouse brain. Predicted reactivity: Mouse (98%), Rat (98%), Zebrafish (85%), Xenopus laevis (96%), Pig (99%), Chicken (98%), Bovine (99%). IHC notes, Requires antigen retrieval using heat mediated 10mM Citrate buffer (pH6.0) or Tris-EDTA buffer (pH8.0) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
v-ets erythroblastosis virus E26 oncogene like isoform 2 (ERG) is a transcriptional regulator. It may participate in transcriptional regulation through the recruitment of SETDB1 histone methyltransferase and subsequent modification of local chromatin structure.
Specifications
| ERG | |
| Polyclonal | |
| Unconjugated | |
| Erg | |
| avian erythroblastosis virus E-26 (v-ets) oncogene related; D030036I24Rik; ERG; ERG, ETS transcription factor; ERG/EWS fusion gene; ERG/FUS fusion gene; ERG/TMPSSR2 fusion gene; ERG1; Erg-3; erythroblast transformation-specific transcription factor ERG variant 10; ETS transcription factor ERG; ets-related; H Kv11.3; HGNC:3446; KCNH2; Oncogene ERG; OTTHUMP00000115970; OTTHUMP00000208036; p55; TMPRSS2/ERG fusion; transcriptional regulator ERG; transcriptional regulator ERG (transforming protein ERG); transcriptional regulator ERG; LOW QUALITY PROTEIN: transcriptional regulator ERG; Transforming protein ERG; v-ets avian erythroblastosis virus E26 oncogene; v-ets avian erythroblastosis virus E26 oncogene homolog; v-ets avian erythroblastosis virus E26 oncogene related; v-ets erythroblastosis virus E26 oncogene homolog; v-ets erythroblastosis virus E26 oncogene like | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13876, 2078 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P11308, P81270 | |
| Erg | |
| Recombinant protein encompassing a sequence within the center region of human ERG. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.