Learn More
Invitrogen™ ERG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588524
Description
Immunogen sequence: MASTIKEALS VVSEDQSLFE CAYGTPHLAK TEMTASSSSD YGQTSKMSPR VPQQDWLSQP PARVTIKMEC NPSQVNGSRN SPDECSVAKG GKMVGSPDTV GMNYGSYMEE KHMPPPNMTT NERRVIVPAD PTLWSTDHVR QWLEWAVKEY GLPDVNILLF QNIDGKELCK MTKDDFQRLT PSYNADILLS HLHYLRETPL PHLTSDDVDK ALQNSPRLMH ARNTGGAAFI FPNTSVYPEA TQRITTRPDL PYEPPRRSAW TGHGHPTPQS KAAQPSPSTV; Positive Samples: MCF7, THP-1, HeLa, BT-474, 293T; Cellular Location: Cytoplasm, Nucleus.
v-ets erythroblastosis virus E26 oncogene like isoform 2 (ERG) is a transcriptional regulator. It may participate in transcriptional regulation through the recruitment of SETDB1 histone methyltransferase and subsequent modification of local chromatin structure.
Specifications
| ERG | |
| Polyclonal | |
| Unconjugated | |
| Erg | |
| avian erythroblastosis virus E-26 (v-ets) oncogene related; D030036I24Rik; ERG; ERG, ETS transcription factor; ERG/EWS fusion gene; ERG/FUS fusion gene; ERG/TMPSSR2 fusion gene; ERG1; Erg-3; erythroblast transformation-specific transcription factor ERG variant 10; ETS transcription factor ERG; ets-related; H Kv11.3; HGNC:3446; KCNH2; Oncogene ERG; OTTHUMP00000115970; OTTHUMP00000208036; p55; TMPRSS2/ERG fusion; transcriptional regulator ERG; transcriptional regulator ERG (transforming protein ERG); transcriptional regulator ERG; LOW QUALITY PROTEIN: transcriptional regulator ERG; Transforming protein ERG; v-ets avian erythroblastosis virus E26 oncogene; v-ets avian erythroblastosis virus E26 oncogene homolog; v-ets avian erythroblastosis virus E26 oncogene related; v-ets erythroblastosis virus E26 oncogene homolog; v-ets erythroblastosis virus E26 oncogene like | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2078 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.54 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P11308 | |
| Erg | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human ERG (NP_8915481). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.