missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERG Mouse anti-Human, Clone: ERG/2107, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07172-20UG
This item is not returnable.
View return policy
Description
ERG Monoclonal specifically detects ERG in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.
Specifications
| ERG | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| transcriptional regulator ERG, 10ets-related, erg-3, p55, TMPRSS2-ERG prostate cancer specific, transcriptional regulator ERG, Transforming protein ERG, v-ets erythroblastosis virus E26 oncogene homolog (avian), v-ets erythroblastosis virus E26 oncogene like, v-ets erythroblastosis virus E26 oncogene like (avian) | |
| Recombinant human full-length ERG protein | |
| 20 μg | |
| Cancer | |
| 2078 | |
| Store at 4°C. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin), Peptide Array | |
| ERG/2107 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction