missing translation for 'onlineSavingsMsg'
Learn More

ERG Antibody (rERG/6843), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18376805 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18376805 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18376805 Supplier Novus Biologicals Supplier No. NBP314204AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ERG Monoclonal antibody specifically detects ERG in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERG
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rERG/6843
Conjugate Alexa Fluor 594
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias transcriptional regulator ERG, 10ets-related, erg-3, p55, TMPRSS2-ERG prostate cancer specific, transcriptional regulator ERG, Transforming protein ERG, v-ets erythroblastosis virus E26 oncogene homolog (avian), v-ets erythroblastosis virus E26 oncogene like, v-ets erythroblastosis virus E26 oncogene like (avian)
Host Species Mouse
Immunogen Recombinant fragment (around aa450 to 479) (C-terminus) of human ERG (exact sequence is proprietary) (Uniprot: P11308)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2078
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.