missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ERG Monoclonal antibody specifically detects ERG in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
Specifications
Specifications
| Antigen | ERG |
| Applications | Immunohistochemistry (Paraffin), Protein Array |
| Classification | Monoclonal |
| Clone | ERG/2107 |
| Conjugate | PerCP |
| Formulation | PBS |
| Gene Alias | transcriptional regulator ERG, 10ets-related, erg-3, p55, TMPRSS2-ERG prostate cancer specific, transcriptional regulator ERG, Transforming protein ERG, v-ets erythroblastosis virus E26 oncogene homolog (avian), v-ets erythroblastosis virus E26 oncogene like, v-ets erythroblastosis virus E26 oncogene like (avian) |
| Host Species | Mouse |
| Immunogen | Recombinant human full-length ERG protein (Uniprot: P11308 ) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?