missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Erfonrilimab Humanized Recombinant Human Monoclonal Antibody

Human Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA554889

Product Code. 30131490

  • £450.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Compatible with Direct ELISA.

Erfonrilimab is a bispecific antibody that targets both PD-1 and CTLA-4, enhancing T-cell activation and anti-tumor immune response.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Erfonrilimab Humanized
Recombinant Monoclonal
Unconjugated
KN 046
Recombinant Human CD274/PD-L1/B7-H1 and CD152/CTLA4.
100 μg
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA
1 mg/mL
PBS with no preservative
Human
Metal-chelate chromatography, Protein A/G
RUO
Human
Antibody
VH-VH'-CH (IgG1)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.