missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eRF1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15765871 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15765871 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15765871 Supplier Invitrogen™ Supplier No. PA557982

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: ILYLTPEQEK DKSHFTDKET GQEHELIESM PLLEWFANNY KKFGATLEIV TDKSQEGSQF VKGFGGIGGI LRYRVDFQGM EYQGGDDEF Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Termination of protein biosynthesis and release of the nascent polypeptide chain are signaled by the presence of an in-frame stop codon at the aminoacyl site of the ribosome. The process of translation termination is universal and is mediated by protein release factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eRF1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ETF1
Gene Accession No. P62495
Gene Alias AI463371; D5S1995; D6Ertd109e; ERF; ERF1; ETF1; Eukaryotic peptide chain release factor subunit 1; eukaryotic release factor 1; eukaryotic translation termination factor 1; polypeptide chain release factor 1; protein Cl1; RF1; sup45 (yeast omnipotent suppressor 45) homolog-like 1; SUP45L1; TB3-1
Gene Symbols ETF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human eRF1. Recombinant protein control fragment (Product #RP-102771).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2107
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.