missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ERF Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530237
This item is not returnable.
View return policy
Description
Recommended positive controls: A431, ERF-transfected 293T. Predicted reactivity: Mouse (99%), Rat (99%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Members of the ETS family of transcription factors, such as ERF, regulate cell proliferation and differentiation. They share a highly conserved DNA-binding domain, the ETS domain, that recognizes the sequence GGAA/T (de Castro et al., 1997 [PubMed 9192842]). For further information on ETS transcription factors, see ETS1 (MIM 164720).
Specifications
| ERF | |
| Polyclonal | |
| Unconjugated | |
| ERF | |
| CRS4; ERF; Est2 repressor factor; ETS domain-containing transcription factor ERF; ETS domain-containing transcription factor ERF; ets2 repressor factor; Ets2 repressor factor; fc22h01; LOW QUALITY PROTEIN: ETS domain-containing transcription factor ERF; PE 2; PE2; PE-2; wu:fc22h01 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13875, 2077 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.24 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P50548, P70459 | |
| ERF | |
| Recombinant protein encompassing a sequence within the center region of human ERF. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction