missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-88120
This item is not returnable.
View return policy
Description
ERF Polyclonal specifically detects ERF in Human, Mouse samples. It is validated for Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Western Blot (Negative).
Specifications
| ERF | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| Ets2 repressor factorPE-2PE2ETS domain-containing transcription factor ERF | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
| 1.0 mg/mL | |
| Immunohistochemistry 1:1000 - 1:5000, Immunoprecipitation 2 - 5 μg/mg lysate, Immunohistochemistry-Paraffin 1:1000 - 1:5000, Western Blot (Negative) | |
| NP_006485.2 | |
| ERF | |
| The immunogen recognized by this antibody maps to a region between residue 498 and 548 of human Ets2 repressor factor using the numbering given in entry NP_006485.2 (GeneID 2077). | |
| 100 μL | |
| Cell Cycle and Replication | |
| 2077 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction