missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERF Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-88120F
This item is not returnable.
View return policy
Description
ERF Polyclonal antibody specifically detects ERF in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ERF | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Ets2 repressor factorPE-2PE2ETS domain-containing transcription factor ERF | |
| The immunogen recognized by this antibody maps to a region between residue 498 and 548 of human Ets2 repressor factor using the numbering given in entry NP_006485.2 (GeneID 2077). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 2077 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| FITC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction