missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Erenumab Recombinant Monoclonal Antibody

Recombinant Monoclonal Antibody

£397.00 - £1002.00

Specifications

Antigen Erenumab
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30283466
View Documents
Invitrogen™
MA558829
100 μg
£397.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30282901
View Documents
Invitrogen™
MA558830
1 mg
£1002.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.

Erenumab (AMG-334) (INN; trade name Aimovig) is a human monoclonal antibody designed specifically to bind and antagonize the calcitonin gene-related peptide receptor (CGRPR) as a means to prevent migraines. Aimovig, as released and marketed by Novartis and Amgen, is in fact a novel therapeutic approach as the first and only FDA approved treatment specifically developed to prevent migraine by blocking the CGRP receptor, which is believed to play a critical role in migraine .
Specifications

Specifications

Erenumab
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Lyophilized
IgG2
Human
AMG 334; erenumab-aooe
Antibody
1 mg/mL
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Unconjugated
Human
RUO
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.