missing translation for 'onlineSavingsMsg'
Learn More

Erdosteine Thioacid Disodium Salt, TRC

Product Code. 30369014
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30369014 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30369014 Supplier Toronto Research Chemicals Supplier No. TRCE596065250MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Erdosteine Thioacid Disodium Salt
CAS 254884-18-9
Purity Grade Notes HPLC
Molecular Formula C8 H11 N O5 S2 . 2 Na
Quantity 250 mg
Synonym N-[[(carboxymethyl)thio]acetyl]homocysteine Disodium Salt (9CI)
InChI Formula InChI=1S/C8H13NO5S2.2Na/c10-6(3-16-4-7(11)12)9-5(1-2-15)8(13)14;;/h5,15H,1-4H2,(H,9,10)(H,11,12)(H,13,14);;/q;2*+1/p-2
SMILES [Na+].[Na+].[O-]C(=O)CSCC(=O)NC(CCS)C(=O)[O-]
IUPAC Name disodium;2-[[2-(carboxylatomethylsulfanyl)acetyl]amino]-4-sulfanylbutanoate
Molecular Weight (g/mol) 311.29
Formula Weight 310.9874
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.