missing translation for 'onlineSavingsMsg'
Learn More

Erdosteine Homocysteine Impurity (Erdosteine RV201), TRC

Product Code. 30388301
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388301 100 mg 100mg
30379667 50 mg 50mg
30369545 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388301 Supplier Toronto Research Chemicals Supplier No. TRCE596070100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Erdosteine Homocysteine Impurity (Erdosteine RV201)
Analyte or Component Names Erdosteine Homocysteine Impurity (Erdosteine RV201)
Shipping Condition Room Temperature
Molecular Formula C12H18N2O5S3
Synonym 2-[N-3-[2-(Oxotetrahydro)thienyl]acetimido]-N-(carboxymethylthioacetyl)homcysteine, [2-[[2-Oxo-2-[(tetrahydro-2-oxo-3-thienyl)amino]ethyl]thio]acetyl]homocysteine, Erdosteine RV 201
InChI Formula InChI=1S/C12H18N2O5S3/c15-9(13-7(1-3-20)11(17)18)5-21-6-10(16)14-8-2-4-22-12(8)19/h7-8,20H,1-6H2,(H,13,15)(H,14,16)(H,17,18)
Solution Type Neat
SMILES O=C1C(NC(CSCC(NC(CCS)C(O)=O)=O)=O)CCS1
IUPAC Name (2-((2-oxo-2-((2-oxotetrahydrothiophen-3-yl)amino)ethyl)thio)acetyl)homocysteine
Industry Type Pharmaceutical
Molecular Weight (g/mol) 366.48
Quantity 100 mg
Formula Weight 366.04
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.