missing translation for 'onlineSavingsMsg'
Learn More

Erdosteine Bis-N-(2-Oxo-3-tetrahydrothienyl) Impurity (Erdosteine EP21506), TRC

Product Code. 30383020
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383020 1 g
30385194 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30383020 Supplier Toronto Research Chemicals Supplier No. TRCE5960751G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C12 H16 N2 O4 S3
Molecular Weight (g/mol) 348.46
Synonym Bis-N-(2-Oxo-3-tetrahydrothienyl)thiodiglycolylamide, Erdosteine EP 21506
IUPAC Name 2-[2-oxo-2-[(2-oxotetrahydrothiophen-3-yl)amino]ethyl]sulfanyl-N-(2-oxotetrahydrothiophen-3-yl)acetamide
SMILES O=C1C(NC(CSCC(NC(CCS2)C2=O)=O)=O)CCS1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H16N2O4S3/c15-9(13-7-1-3-20-11(7)17)5-19-6-10(16)14-8-2-4-21-12(8)18/h7-8H,1-6H2,(H,13,15)(H,14,16)
Formula Weight 348.0272
Percent Purity >95%
Chemical Name or Material Erdosteine Bis-N-(2-Oxo-3-tetrahydrothienyl) Impurity
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.