missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ERCC8 Recombinant Rabbit Monoclonal Antibody (23GB5905), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552946
This item is not returnable.
View return policy
Description
This gene encodes a WD repeat protein, which interacts with Cockayne syndrome type B (CSB) protein and with p44 protein, a subunit of the RNA polymerase II transcription factor IIH. Mutations in this gene have been identified in patients with hereditary disease Cockayne syndrome (CS). CS cells are abnormally sensitive to ultraviolet radiation and are defective in the repair of transcriptionally active genes.
Specifications
| ERCC8 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ERCC8 | |
| 2410022P04Rik; 2810431L23Rik; 4631412O06Rik; B130065P18Rik; Ckn1; Cockayne syndrome 1 (classical); Cockayne syndrome 1 homolog; cockayne syndrome WD repeat protein CSA; Cockayne syndrome WD repeat protein CSA homolog; Cockayne syndrome WD-repeat protein CSA; CSA; DNA excision repair protein ERCC-8; ERCC excision repair 8, CSA ubiquitin ligase complex subunit; Ercc8; excision repair cross-complementation group 8; excision repair cross-complementing rodent repair deficiency complementation group 8; excision repair cross-complementing rodent repair deficiency, complementation group 8; excision repaiross-complementing rodent repair deficiency, complementation group 8; UVSS2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1161 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5905 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13216 | |
| ERCC8 | |
| A synthesized peptide derived from human ERCC8 (351-396AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction