missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ERCC8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521281
This item is not returnable.
View return policy
Description
Recommended positive controls: Molt-4, C8D30, Rat2. Predicted reactivity: Mouse (91%), Rat (91%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a WD repeat protein, which interacts with Cockayne syndrome type B (CSB) protein and with p44 protein, a subunit of the RNA polymerase II transcription factor IIH. Mutations in this gene have been identified in patients with hereditary disease Cockayne syndrome (CS). CS cells are abnormally sensitive to ultraviolet radiation and are defective in the repair of transcriptionally active genes.
Specifications
| ERCC8 | |
| Polyclonal | |
| Unconjugated | |
| ERCC8 | |
| 2410022P04Rik; 2810431L23Rik; 4631412O06Rik; B130065P18Rik; Ckn1; Cockayne syndrome 1 (classical); Cockayne syndrome 1 homolog; cockayne syndrome WD repeat protein CSA; Cockayne syndrome WD repeat protein CSA homolog; Cockayne syndrome WD-repeat protein CSA; CSA; DNA excision repair protein ERCC-8; ERCC excision repair 8, CSA ubiquitin ligase complex subunit; Ercc8; excision repair cross-complementation group 8; excision repair cross-complementing rodent repair deficiency complementation group 8; excision repair cross-complementing rodent repair deficiency, complementation group 8; excision repaiross-complementing rodent repair deficiency, complementation group 8; UVSS2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 1161, 310071, 71991 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 5.39 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q13216, Q8CFD5 | |
| ERCC8 | |
| Recombinant protein encompassing a sequence within the center region of human ERCC8. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction