missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ERCC6L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550405
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total ERCC6L protein.
ERCC6L is a DNA helicase that acts as an essential component of the spindle assembly checkpoint. ERCC6L contributes to the mitotic checkpoint by recruiting MAD2 to kinetochores and monitoring tension on centromeric chromatin. ERCC6L acts as a tension sensor that associates with catenated DNA which is stretched under tension until it is resolved during anaphase.
Specifications
| ERCC6L | |
| Polyclonal | |
| Unconjugated | |
| Ercc6l | |
| ATP-dependent helicase ERCC6-like; BC004701; D330021P09Rik; DNA excision repair protein ERCC-6-like; ERCC excision repair 6 like, spindle assembly checkpoint helicase; ERCC6L; excision repair cross-complementation group 6 like; excision repair cross-complementation group 6-like; excision repair cross-complementing rodent repair deficiency complementation group 6 - like; excision repair cross-complementing rodent repair deficiency complementation group 6 like; excision repair cross-complementing rodent repair deficiency complementation group 6-like; excision repair cross-complementing rodent repair deficiency, complementation group 6-like; excision repair protein ERCC6-like; FLJ20105; LOW QUALITY PROTEIN: DNA excision repair protein ERCC-6-like; MGC131695; PICH; PLK1-interacting checkpoint helicase; RAD26L; RGD1565734; SNF2/RAD54 family protein; tumor antigen BJ-HCC-15 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54821 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.2 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q2NKX8 | |
| Ercc6l | |
| Fusion protein corresponding to residues near the C terminal of human excision repair cross-complementation group 6-like. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction