missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERCC6L Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-97609CL1
This item is not returnable.
View return policy
Description
ERCC6L Polyclonal antibody specifically detects ERCC6L in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ERCC6L | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| ATP-dependent helicase ERCC6-like, DNA excision repair protein ERCC-6-like, EC 3.6.1, EC 3.6.4.12, excision repair cross-complementing rodent repair deficiency, complementationgroup 6-like, excision repair protein ERCC6-like, FLJ20105, MGC131695, PICHexcision repair cross-complementing rodent repair deficiency complementationgroup 6 - like, PLK1-interacting checkpoint helicase, SNF2/RAD54 family protein, Tumor antigen BJ-HCC-15 | |
| Produced in rabbits immunized with E. coli-derived Human ERCC6L fragment. | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 54821 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction