missing translation for 'onlineSavingsMsg'
Learn More

ERCC6L Antibody (3F12-2B10), Novus Biologicals™

Product Code. 18366419 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18366419 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18366419 Supplier Novus Biologicals Supplier No. H00054821M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ERCC6L Monoclonal antibody specifically detects ERCC6L in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERCC6L
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 3F12-2B10
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100-1:2000
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH08808
Gene Alias ATP-dependent helicase ERCC6-like, DNA excision repair protein ERCC-6-like, EC 3.6.1, EC 3.6.4.12, excision repair cross-complementing rodent repair deficiency, complementationgroup 6-like, excision repair protein ERCC6-like, FLJ20105, MGC131695, PICHexcision repair cross-complementing rodent repair deficiency complementationgroup 6 - like, PLK1-interacting checkpoint helicase, SNF2/RAD54 family protein, Tumor antigen BJ-HCC-15
Host Species Mouse
Immunogen ERCC6L (AAH08808, 1 a.a. ~ 419 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MEKSFATKNEAVQKETLQEGPKQEALQEDPLESFNYVLSKSTKADIGPNLDQLKDDEILRHCNPWPIISITNESQNAESNVSIIEIADDLSASHSALQDAQASEAKLEEEPSASSPQYACDFNLFLEDSADNRQNFSSQSLEHVEKENSLCGSAPNSRAGFVHSKTCLSWEFSEKDDEPEEVVVKAKIRSKARRIVSDGEDEDDSFKDTSSINPFNTSLFQFSSVKQFDASTPKNDISPPGRFFSSQIPSSVNKSMNSRRSLASRRSLINMVLDHVEDMEERLDDSSEAKGPEDYPEEGVEESSGEASKYTEEDPSGETLSSENKSSWLMTSKPSALAQETSLGAPEPLSGEQLVGSPQDKAAEATNDYETLVKRGKELKECGKIQEALNCLVKALDIKSADPEVMLLTLSLYKQLNNN
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 54821
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.