missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERCC2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10818-1-AP-20UL
This item is not returnable.
View return policy
Description
XPD is involved with the nucleotide excision repair pathway that is a mechanism to repair damage to DNA. The protein encoded by this gene is involved in transcription-coupled nucleotide excision repair and is an integral member of the basal transcription factor BTF2/TFIIH complex. The gene product has ATP-dependent DNA helicase activity and belongs to the RAD3/XPD subfamily of helicases. Defects in this gene can result in three different disorders, the cancer-prone syndrome xeroderma pigmentosum complementation group D, trichothiodystrophy, and Cockayne syndrome. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| ERCC2 | |
| Polyclonal | |
| Unconjugated | |
| ERCC2 | |
| BTF2 p80, COFS2, CXPD, EM9, ERCC2, TFIIH 80 kDa subunit, TFIIH p80, TTD, XPD, XPDC | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2068 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P18074 | |
| ERCC2 | |
| ERCC2 Fusion Protein Ag1038 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction