missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ERCC1 Recombinant Rabbit Monoclonal Antibody (23GB1230), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA554189
This item is not returnable.
View return policy
Description
ERCC1 is a non-catalytic component of a structure-specific DNA repair endonuclease responsible for the 5'-incision during DNA repair. It is responsible, in conjunction with SLX4, for the first step in the repair of interstrand cross-links (ICL). It participates in the processing of anaphase bridge-generating DNA structures, which consist in incompletely processed DNA lesions arising during S or G2 phase, and can result in cytokinesis failure. ERCC1 is also required for homology-directed repair (HDR) of DNA double-strand breaks, in conjunction with SLX4. (Uniprot).
Specifications
| ERCC1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ERCC1 | |
| COFS4; DNA excision repair protein ERCC-1; ERCC excision repair 1, endonuclease non-catalytic subunit; Ercc1; Ercc-1; excision repair cross-complementation group 1; excision repair cross-complementing 1; excision repair cross-complementing rodent repair deficiency, complementation group 1; excision repair cross-complementing rodent repair deficiency, complementation group 1 (includes overlapping antisense sequence); hypothetical protein LOC405807; RAD10; UV20; zgc:77511 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13870, 2067 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB1230 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P07903, P07992 | |
| ERCC1 | |
| A synthesized peptide derived from human ERCC1 (1-180AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction